HG-200 GenoLyte® Compact Tissue Homogenizer and Cell Lyser
Supplier: SPEX
1200
76324-998EA 5819.76 USD
76324-998
HG-200 GenoLyte® Compact Tissue Homogenizer and Cell Lyser
Homogenizers Benchtop Homogenizers
Ideal for small-scale sample preparation.
  • High-intensity oscillating motion homogenizes samples in seconds
  • Keep an eye on the grind process with the clear lid for viewing
  • Interchangeable vial holders handle a range of vial sizes from 2 to 12 ml
  • Safety interlock switch stops the motor if the lid is opened during homogenization
  • Digital display screen with rate and timer controls allows you to adjust speed and set time
The HG-200 GenoLyte® is capable of blending powders and crushing rocks to a fine powder or DNA, RNA, and protein extractions. It is specifically designed for rapid cell disruption, cell lysis and tissue homogenization. Common samples include plant material, seeds, animal tissue, bacteria, yeast, stool, and soil. This homogenizer is compact yet powerful, which frees up bench space in the lab. Homogenizer accepts a vial holder for 2 to 12 ml vials, a jar holder for 75 ml jars, or titer plates (all sold separately).