Tronex® 9117 Series NEW AGE® Chemo-Rated Nitrile Examination Gloves, Tronex International Inc.
Supplier: Tronex
9117-10
76324-964BX 71.73 USD
76324-964 76324-974 76324-976 76324-978
Tronex® 9117 Series NEW AGE® Chemo-Rated Nitrile Examination Gloves, Tronex International Inc.
Gloves
Tronex® nitrile exam-grade gloves feature an ambidextrous fit with beaded cuffs. They are designed to meet or exceed all industry standards for safety and quality.
  • Excellent barrier protection, comfort, and chemical resistance
  • Fingertip textured for improved grip
  • Not made with natural rubber latex, powder-free, non sterile
Tronex® 9117 series examination gloves feature an innovative nitrile material formulation for exceptional fit and feel properties for applications with shorter durations. Gloves offer excellent barrier protection for bloodborne pathogens, as well as resistance to chemotherapy drugs and other chemicals.
Certifications: Chemo-rated per ASTM D6978.
Packaging: Gloves are packaged 250 per box, 10 boxes per case.