Whatman™ Mini-UniPrep MultiCompressor, Whatman products (Cytiva)
Supplier: Cytiva
MUPMCPBC8
76324-940EA 456.67 USD
76324-940
Whatman™ Mini-UniPrep MultiCompressor, Whatman products (Cytiva)
Presses
Mini-UniPrep™ multicompressor including one 8-position tray for standard plastic Mini-UniPrep™ filter vials.
  • Streamlines syringeless filtration by filtering up to eight filter vials simultaneously
  • Designed for Cytiva's Mini-UniPrep™ standard plastic filter vial
  • Mini-UniPrep™ vials can be loaded directly into the autosampler
The newly redesigned Whatman™ Mini-UniPrep™ multicompressor streamlines filtration, saves time and reduces hand stress by enabling the filtration of eight Mini-UniPrep™ vials at once.
Ordering information: Additional trays are available, giving the added convenience of pre-loading up to eight Mini-UniPrep™ filter vials.
Delivery information: The multiprocessor includes one blue 8-position tray.