VWR® LTS Compatible Pipette Tips
Supplier: VWR International
76322-158 76322-160 76322-162 76322-164 76322-172 76322-174 76322-176 76322-178 76322-180 76322-530 76322-532 76322-534 76323-932 76323-934 76323-936 76323-938 76323-940 76323-942 76323-944 76323-946 76327-210 76327-212 76327-216 76327-218 76327-220
76322-158CS 589.43 USD
76322-158 76322-160 76322-162 76322-164 76322-172 76322-174 76322-176 76322-178 76322-180 76322-530 76322-532 76322-534 76323-932 76323-934 76323-936 76323-938 76323-940 76323-942 76323-944 76323-946 76327-210 76327-212 76327-216 76327-218 76327-220
VWR® LTS Compatible Pipette Tips
Pipette Tips
VWR® LTS™ Compatible pipette tips feature a flexible seal area and a positive stop resulting in a secure seal with light ejection force on LTS™ compatible pipettors.
  • Compatible with the LTS™ style pipettors
  • Maximizes sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Autoclavable at 15 psi, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than otherMore Product Information