VWR® Universal Pipette Tips
Supplier: VWR International
63305017 63305018 63305019 63305030 63305031 63305032 63305033 63305034 63305035 63305020 63305021 63305022 63305023 63305046 63305047 63305048 63305045 63305038 63305039 63305040 63305041 63305042 63305049 63305050 63305051 63305024 63305025 63305026 63305027 63305028 63305029 63305036 63305037 63305016 63305054 63305373 63305368 63305369 63305374 63305370 63305371 63305372
76322-132CS 796.11 USD
76322-132 76322-134 76322-136 76322-138 76322-140 76322-142 76322-144 76322-146 76322-148 76322-150 76322-152 76322-154 76322-156 76323-388 76323-390 76323-392 76323-394 76323-396 76323-398 76323-400 76323-402 76323-404 76323-452 76323-454 76323-456 76322-512 76322-514 76322-516 76322-518 76322-520 76322-522 76322-524 76322-526 76322-528 76327-214 76595-790 76595-792 76595-798 76595-788 76595-794 76595-800 76595-796
VWR® Universal Pipette Tips
Pipette Tips
VWR® Universal low retention pipette tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.
  • Compatible with a wide range of pipettes
  • Maximizes sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Autoclavable at 15 psi, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropyleneMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Sustainable Packaging