VWR® Universal Pipette Tips
Supplier: VWR International
76322-132 76322-134 76322-136 76322-138 76322-140 76322-142 76322-144 76322-146 76322-148 76322-150 76322-152 76322-154 76322-156 76323-388 76323-390 76323-392 76323-394 76323-396 76323-398 76323-400 76323-402 76323-404 76323-452 76323-454 76323-456 76322-512 76322-514 76322-516 76322-518 76322-520 76322-522 76322-524 76322-526 76322-528 76327-214 76595-790 76595-792 76595-798 76595-788 76595-794 76595-800 76595-796
76322-132CS 796.11 USD
76322-132 76322-134 76322-136 76322-138 76322-140 76322-142 76322-144 76322-146 76322-148 76322-150 76322-152 76322-154 76322-156 76323-388 76323-390 76323-392 76323-394 76323-396 76323-398 76323-400 76323-402 76323-404 76323-452 76323-454 76323-456 76322-512 76322-514 76322-516 76322-518 76322-520 76322-522 76322-524 76322-526 76322-528 76327-214 76595-790 76595-792 76595-798 76595-788 76595-794 76595-800 76595-796
VWR® Universal Pipette Tips
Pipette Tips
VWR® Universal low retention pipette tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.
  • Compatible with a wide range of pipettes
  • Maximizes sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Autoclavable at 15 psi, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropyleneMore Product Information