Transparent Semi-Permeable Dressings, DUKAL Corporation®
Supplier: Dukal
4067
76321-918BX 133.71 USD
76321-918
Transparent Semi-Permeable Dressings, DUKAL Corporation®
First Aid Supplies Wound Dressings
Ideal for IV sites, minor burns, ulcers, skin tears, and donor sites.
  • Easy two-step application
  • Protects against fluid contaminants and bacteria
  • Rounded edges prevent roll up
  • Removable patient label for chart
  • Not made with natural rubber latex
Microporous transparent film offers a high Moisture Vapor Transmission Rate (MVTR).
Packaging: Dressings are packaged 1 per pack, 100 packs per box.