Mag-Bind® Total RNA 96 Kit
Supplier: OMEGA BIO-TEK
M6731-00
76321-016EA 408.12 USD
76321-016
Mag-Bind® Total RNA 96 Kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Isolate total cellular RNA and viral RNA from a wide variety of cells and tissues.
  • Procedure can be fully automated with most robotic workstations
The Mag-Bind® Total RNA 96 Kit provides a novel technology for total RNA isolation. This kit allows the rapid and reliable isolation of high-quality total cellular RNA and viral RNA from a wide variety of cells and tissues. Unlike column-based systems, the binding of nucleic acids to magnetic particles occurs in solution resulting in increased binding kinetics and binding efficiency. Particles are also completely re-suspended during the wash steps of the purification protocol, which improves the removal of contaminants and increases nucleic acid purity. Mag-Bind® Total RNA 96 Kit procedure can be fully automated with most robotic workstations.