Cuvette Swabs, 6" Spatula Foam Swab with Black Polypropylene Handle, Hardy Diagnostics
Supplier: Hardy Diagnostics
QS1001
76320-912PK 67.96 USD
76320-912
Cuvette Swabs, 6" Spatula Foam Swab with Black Polypropylene Handle, Hardy Diagnostics
Swabs Swabs for Sampling
Specifically designed for QuickSlide™ automated stainers, the foam-tipped swab is ideal for maintaining a clean cuvette, free of stain residue. Foam-tipped swabs are a necessity as cotton swabs leave behind lint that can potentially cause an overflow in the cuvette.
  • Helps maintain a clean, lint-free cuvette
  • Improves decolorization accuracy
  • Foam tip absorbs residue quickly
  • Quick five-minute procedure
  • 100 swabs per pack