Media/Packaging Bottles, PETG, Square, TriForest
Supplier: TRIFOREST ENTERPRISES INC
PGC2000MS-C PGC1000MS-C PGC0500MS-A PGC0500MS-C PGC0250MS-A PGC0250MS-C PGC0125MS-A PGC0125MS-C PGC0060MS-C PGC0030MS-C
76320-364CS 246.59 USD
76320-364 76320-366 76320-368 76320-370 76320-372 76320-374 76320-376 76320-378 76320-380 76320-382 76320-384 76320-386 76320-388 76320-390 76320-392 76320-394 76320-396 76320-398 76320-400 76320-402 76320-404
Media/Packaging Bottles, PETG, Square, TriForest
Bottles Plastic Bottles
TriForest Square PETG bottles are ideal for storage of media, reagents, and buffers.
  • Thick-walled and durable; shatter resistant
  • Excellent gas barrier properties; reduced permeability to CO2/O2
  • Sterility of 10⁻⁶ SAL
  • Molded graduations for approximate volume reference
  • Compliant with USP Class VI guidelines
  • Non-pyrogenic, nuclease-free, and non-cytotoxic
  • Bottle and cap material are Certified Animal Derived Component-Free
  • Two litre bottles have a unique design with molded-in hand/robotic grips
Sterile polyethylene terephthalate co-polyester bottles are available with or without high density polyethylene caps in shrink wrapped trays. These heavy-duty, break-resistant bottles are designed for manual and automated fill lines.
Bottles are supplied shrink wrapped in polymer trays to eliminate static adherence to particulate matters. Double poly-bagged for unwrapping in cleanroom operations to maintain the utmost integrity of cleanroom based sterile production/filling/packaging requirements.
Bottles are available with standard cap (PGCXXXXMS-C), tamper evident cap (PGCXXXXMS-A), or without cap (PGCXXXXMS-N) for use with automated fill lines.