xPIPETTE™ Carousel Stand
Supplier: Biotix
30518126
76320-254CS 100.92 USD
76320-254
xPIPETTE™ Carousel Stand
Pipette Stands
The carousel pipette stand is a rotating carousel stand for the xPIPETTE.
  • Fits both single and multichannel models
  • Holds seven pipettes total
  • Made from chemically resistant polypropylene
Pipettes hang vertically, preventing liquids from contaminating the pipette shaft.