Purifier® Logic®+ Class II B2 Biosafety Cabinets, Labconco
Supplier: Labconco
303481151
76319-916EA 22080 USD
76319-916 76319-918 76319-920 76319-922 76319-924 76319-926 76319-928 76319-930 76319-932 76319-934 76319-936 76319-938 76319-940 76319-942 76319-944 76319-946 76319-948 76319-950 76319-952 76319-954 76319-956 76319-958 76319-960 76319-962 76319-964 76319-966 76319-968 76319-970 76319-972 76319-974 76319-976 76319-978 76319-980 76319-982 76319-984 76319-986 76319-988 76319-990 76319-992 76319-994 76319-996 76319-998 76320-000 76320-002 76320-004 76320-006 76320-008 76320-010 76320-036 76320-038 76320-040 76320-042 76320-044 76320-046 76320-048 76320-050
Purifier® Logic®+ Class II B2 Biosafety Cabinets, Labconco
Fume Hoods and Enclosures Biological Safety Cabinets
Purifier® Logic®+ type B2 biosafety cabinets combine ergonomics, comfort and safety.
  • MyLogic OS displays cabinet status and alarms with clear, concise images and messaging to build user confidence
  • Constant Airflow Profile™ (CAP) Technology automatically maintains a precise volume of air as the filters load to protect a user and their samples
  • Ergonomic Inclination™ Technology Suite includes 10° angled sash, patented curved air grille with built-in armrest, fully accessible controls and eye-level mounted color display
  • Quiet, efficient ECM blower and energy saving LED lights reduces cabinet’s carbon footprint
  • 100% exhaust design provides personnel, product and environmental protection and is suitable for work in all biosafety levels
The Purifier Logic+ Class II B2 Biosafety Cabinet excels in user comfort and functionality. Decades of industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, LED lighting, patented curved air grille with built in armrest, fully accessible controls and line-of-sight mounted color display with MyLogic™ operating system.
MyLogic OS provides intuitive control of the Logic+ with clear messages and vibrant imagery, status updates and alarms with diagnostic troubleshooting. Using ECM blower technology, the Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
Type B2 biosafety cabinets exhaust 100% of their air and are designed to be hard ducted to the outside. These cabinets provide personnel, product and environmental protection and are suitable for work in all biosafety levels. In addition, some models have base stands and/or an Accessory Package (UV light, service fixture(s) and Vacu-Pass™ Cord and Cable Portal) included.
Certifications: ETL listed (North America and Japan only); NSF international listed; CE conformity marking (230 V models); ISO 5 conditions per ISO 14644-1 and 2; ADA-compliant (controls and receptacles).
Accessories information: Accessories not included unless stated.
Ordering information: Base stand required if model with stand is not selected.