Micro Spatula, Stainless Steel, Bürkle
Supplier: VWR International
5386-0001
76319-752EA 7.95 USD
76319-752
Micro Spatula, Stainless Steel, Bürkle
Spatulas
The micro spatula can be used for reliably measuring, sampling, or processing small amounts of powder, chemicals, pastes, creams, or active ingredients.
  • Stainless steel AISI 304 (1.4301)
  • High-gloss polished
  • Manufactured entirely without edges, crevices, or undercuts
This eliminates the possibility of impurities or cross-contamination, as no product residue or cleaning agents can be deposited. The micro spatula is ideal for use in the pharmaceutical industry as well as food and beverage, and cosmetics.