One-Piece Glass Reaction Vessel, 5-Neck, 100 ml, Chemglass
Supplier: Chemglass
CG-MR-100V1P
76319-656EA 650.7 USD
76319-656
One-Piece Glass Reaction Vessel, 5-Neck, 100 ml, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
One-piece reaction vessel with five necks features: 19/22 Outer CN, (1) 10/18 Outer SN, (1) 14/20 Outer SN, and (2) 19/22 Outer SN's.
  • Vessel is graduated from 20 to 100 ml
  • Body outside diameter is 57 mm
  • Equivalent to Mettler Easymax®
Equivalent to Mettler EasyMax 102/402 Part # 51161786. Mettler Easymax® is a registered trademark of Mettler Toledo.