Sakura Prisma H&E Stain Kit
Supplier: SAKURA FINETEK USA INC
6190
76318-762KT 490.19 USD
76318-762
Sakura Prisma H&E Stain Kit
Stains Histology Stains
Validated, optimized H&E staining protocol for the Prisma automatic slide stainer.
  • Patented, custom-designed bottle with ergonomic grips make easy to handle and less spills
  • Each kit and component are barcoded to enable scanning of the lot numbers and expire date for documentation on Prisma Plus automatic stainer
  • Biological stain commission-certified dyes are use in the manufacturing process to ensure highest quality components
  • Single kit will stain up to 2,500 slides and last up to two weeks
  • Color-coded labels and bottle caps which match the colors of the solution configuration of the Prisma Plus stainer and avoices filling errors
  • The 750 ml fill size, ready-to-use bottles are designed to match the recommended volume for standard Prisma stainer solution reservoirs
The ready to use kit consists of 6 bottles (1 each hematoxylin and eosin, 2 each differentiator and bluing reagent) eliminating the need to measure or dilute reagents.
Shelf life: Unopened 18 months from date of manufacture, once opened 14 days.
Caution: The product can expose you to chemicals including Ethylene Glycol and Methanol.