Diesse Cube 30 Touch, Streck
Supplier: Streck Laboratories
240418
76318-736EA 14157.5 USD
76318-736
Diesse Cube 30 Touch, Streck
Clinical Diagnostic Supplies Clinical Analysers
An automated instrument for high-volume erythrocyte sedmentation rate testing in EDTA tubes.
  • 94% correlation to modified Westergren method
  • Compatible with 13×75 mm (1.5 to 4.0 ml) K2EDTA or K3EDTA tubes
  • Results in 20 minutes
  • 30 Samples/Batch
  • Internal barcode scanner and printer
  • Automated patient and QC data archive
  • Easily interfaces to LIS
  • No reagents, no exposure to patient sample