ARM-D Kit, β-Lactamase, Steck
Supplier: STRECK LABORATORIES
250045
76318-696EA 2977.34 USD
76318-696
ARM-D Kit, β-Lactamase, Steck
Assays qPCR Assays
The Streck ARM-D Kit, β-Lactamase, is a comprehensive, TaqMan based method for the detection of antibiotic resistance gene families in Gram Negative Bacteria, including the five CRE's described by the CDC as urgent public health threats.
  • Detects over 450 gene variants
  • Dectects the five major CRE resistance families
  • Dectects ESBL's and ampC resistance markers
  • Understand resistance mechanisms prior to obtaining susceptibility results
  • Works on a large variety of instrumentation