Radiance ECL, Azure Biosystems
Supplier: Azure Biosystems
AC2204
76318-638EA 451.37 USD
76318-638
Radiance ECL, Azure Biosystems
Electrophoresis Reagents Western Blotting Reagents
Radiance ECL is an HRP substrate with long lasting signal and sensitivity down to low picograms. Use up to 10 times less antibody than other ECL substrates.
  • Compare performance to SuperSignal West Pico and Amersham ECL
  • Sensitivity in low picograms
  • Signal duration 6 to 8 hours