MicroIntensive™ L Sleeve, Vileda Professional®
Supplier: Vileda Professional - FHP
162386
76318-348CS 1018.42 USD
76318-348
MicroIntensive™ L Sleeve, Vileda Professional®
Mops
Single use, double-sided Microfiber blend sleeve, is pre-laundered and single bagged. With excellent cleaning performance and high absorbency it is ideal for pre-prepared methods or pull and lift. Independently proven bacteria reduction of 99.99%.
  • Sleeve fits CE MultiSurface Cleaner™
  • Single use and double sided
  • Pre-Laundered, single bagged, and lot # controlled
  • Four layer construction
  • High absorbency with excellent cleaning performance
Customized tools for special surfaces. The easiest, most effective way to clean pipes, soft walls curtains, tanks and other hard to reach areas in Controlled Environments.
A new line of specialty above-the-floor tools by Vileda Professional®. This line is ideal for all surfaces that are hard or difficult to reach, clean, and disinfect. Specialty tools use Controlled Environment compatible disposable microfiber sleeves that are already validated materials and are an extension of a current CE floor cleaning system.
Certifications: Meet CE requirements up to ISO 4-9/GMP A/B, C, D.