CE Tank Cleaner™, Cleaning Mop, Vileda Professional®
Supplier: Vileda Professional - FHP
162234
76318-340CS 695.07 USD
76318-340
CE Tank Cleaner™, Cleaning Mop, Vileda Professional®
Mops
CE Tank Cleaner™ is designed from electro-polished stainless steel and is pre-bent to a 9" radius for an easily adjustable fit into inner or outer tank walls. This unit is easy to handle for tight spaces, or can be used with the telescopic handle for hard to reach areas.
  • Ideal for inner and outer tanks walls
  • Completely autoclavable
  • Frame is pre-bent for tanks, but can also be used flat for isolators
  • Four degrees of freedom to ensure good contact with surface being cleaned
Customized tools for special surfaces. The easiest, most effective way to clean pipes, soft walls curtains, tanks and other hard to reach areas in Controlled Environments.
A new line of specialty above-the-floor tools by Vileda Professional®. This line is ideal for all surfaces that are hard or difficult to reach, clean, and disinfect. Specialty tools use Controlled Environment compatible disposable microfiber sleeves that are already validated materials and are an extension of a current CE floor cleaning system.
Certifications: Meet CE requirements up to ISO 4-9/GMP A/B, C, D.