MX350 Protective Eyewear, HexArmor
Supplier: HexArmor
11-23001-04 11-23002-04 11-23003-04 11-23004-04
76317-752CS 783.4 USD
76317-752 76317-754 76317-756 76318-172
MX350 Protective Eyewear, HexArmor
Safety Glasses
The MX350 series gasketed eyewear features an adjustable head strap, innovative soft seal frame technology with brow guards, and a soft nosepiece for premium comfort, protection, and optical clarity.
  • Soft seal frame technology adapts to the face for a customized fit
  • Innovative brow guard provides additional protection from flying particles, dirt, or dust
  • LTX® lens technology for premium optical clarity
  • 100% UV protection
Sleek MX350 protective eyewear offers full-coverage gasketed eyewear to keep dust and debris out of the eyes. Flex technology and a soft nosepiece deliver a low-pressure soft seal that adapts to the face, while providing D3/D4 liquid splash-rated ventilation. Available in multiple tints, the fully adjustable head strap gives wearers a secure, pressure-free fit (available in cloth and gas-resistant neoprene), ideal for multiple job tasks.
Certifications: Compliant with ANSI Z87.1.2015. High Impact Rated Z87+. CE EN 166