Helix® 2088 Series Gloves, Hexarmor
Supplier: HexArmor
2088-XS(6) 2088-XXXL(12)
76317-716CS 607.58 USD
76317-716 76317-718 76317-720 76317-722 76317-724 76317-726 76317-728 76317-730
Helix® 2088 Series Gloves, Hexarmor
Gloves
Seamless, coated Helix® series gloves feature a 13-gauge, HPPE and fiberglass blend shell offering 360° cut protection with ANSI/ISEA level A4 cut resistance. A flexible foam nitrile three-quarter palm and finger coating provides superior grip and abrasion resistance.
  • Flexible foam nitrile ³/₄ palm and finger coating provides extra coverage and superior grip and abrasion resistance
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Silicone-free
  • REACH Compliant for being void of harsh chemicals
Helix® series gloves offer a wide variety of grip options to provide users with the appropriate grip for the task at hand. Providing coverage for any working condition, HexArmor is constantly researching and testing new formulas and application methods to maximize grip and function, helping workers redefine safety at their worksite.
Certifications: EN 388. CE 4543. ANSI/ISEA Cut A4. REACH Compliant.