Large Scissors, Rounded Tip, Slice®
Supplier: SLICE, INC.
10545
76314-906CS 398.93 USD
76314-906
Large Scissors, Rounded Tip, Slice®
Scissors
Cut through tough materials like Kevlar-reinforced fabric with Slice® safety scissors. These rounded tip scissors feature comfortable soft-touch grips and handles large enough for gloved hands.
  • Zirconium oxide Slice® safety blades
  • Durable reinforced nylon handle with soft-touch finger grips
  • Safer than traditional scissors
  • Finger-friendly® edge lasts 11 times longer than metal
  • Ambidextrous design
  • Reduces injuries, lower costs
  • Non-sparking, non-conductive, non-magnetic blades
  • Chemically inert blades never rust
  • Oil, lubricant, and maintenance free
  • BPA-, phthalate-, and lead-free scissors
  • Lightweight and compact
The 100% zirconium oxide blades are finger-friendly® for added safety, and last up to 11 times longer than comparable steel blades. These BPA- and lead-free scissors are strong enough for adults and safe enough for kids to use. Chemically inert, oil-free, and non-rusting, the 60mm blades of these lightweight scissors are easy to clean and require no maintenance.