Manual Utility Scraper with Safety Blade, Slice®
Supplier: SLICE, INC.
10591
76314-896CS 394.07 USD
76314-896
Manual Utility Scraper with Safety Blade, Slice®
Scrapers
Slice® pairs safety and utility with the Manual Utility Scraper, featuring the patent-pending finger-friendly® safety blade.
  • Installed blade type: rounded tip
  • Durable nylon handle with soft-touch comfort grips
  • Ambidextrous design
  • Compatible blades: 10526, 10528
  • Finger-friendly® edge lasts 11 times longer than metal
  • Chemically inert blade never rusts
  • Non-sparking, non-conductive, non-magnetic blade
  • Oil- and lubricant-free blade
  • Reduces injuries, lowers costs
  • No-tool blade change
This unique Slice® design creates a blade that’s safe to the touch and much more durable than traditional blades. In addition, the 100% zirconium oxide blade is non-conductive, chemically inert, and never rusts. The scraper’s handle is ergonomically designed with soft-touch finger grips for extra comfort and includes convenient touches like a built-in lanyard hole and tool-free blade change. This ambidextrous scraper is equipped with a rounded-tip utility blade, ideal for scraping paint, rust, and adhesives.