VWR® MINI LR Horizontal Gel Electrophoresis System
Supplier: VWR International
80-1214R
76314-796EA 1385.7 USD
76314-796
VWR® MINI LR Horizontal Gel Electrophoresis System
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The MINI LR model includes a buffer circulation system, powered by the electrodes themselves.
In constrast to all the other Mini gel systems, the MINI LR model includes a buffer circulation system, powered by the electrodes themselves. Hydrogen bubbles produced at the cathode pass up a diagonally positioned tube connected to the two sides of the tank producing a gentle circulation of the buffer. The result is a no pH and ionic gradient - without the need of mechanical pumps.
Delivery information: Includes integral buffer circulation, safety lid with retractable sheathed leads, UVT gel tray, two combs, one 12 well and one 20 well (1.5 mm thick).