VWR® Horizontal MINI L Gel Electrophoresis System
Supplier: VWR International
80-1214
76314-778EA 956.11 USD
76314-778
VWR® Horizontal MINI L Gel Electrophoresis System
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The MINI L is the largest of the 'Mini-Gel' range is supplied with a four slot UVT gel tray capable of running 100 samples.
There is also a 12 slot tray available. There is an expansive range of standard and MC-compatible loading combs, making this versatile system appropriate for both routine and small separation electrophoresis.
Delivery information: Includes a safety lid with retractable sheathed leads, UVT gel tray, two combs, one 12 well and one 20 well (1.5 mm thick).