VWR® Horizontal MINI M Gel Electrophorosis Systems
Supplier: VWR International
80-0911
76314-748EA 887.08 USD
76314-748
VWR® Horizontal MINI M Gel Electrophorosis Systems
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The MINI M electrophoresis system is a very convenient size for a wide range of routine uses. A wide range of standard and MC-compatible loading combs are available, separation of up to 36 samples.
Delivery information: Includes a safety lid with retractable sheathed leads, UVT gel tray and two combs, one each 10 well and 14 well (1.5 mm thick).