VWR® Horizontal MAXI L Gel Electrophoresis System with Quick-Disconnect Buffer Exchange Ports
Supplier: VWR International
81-2340BEP
76314-704EA 1779.24 USD
76314-704
VWR® Horizontal MAXI L Gel Electrophoresis System with Quick-Disconnect Buffer Exchange Ports
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Delivery information: Includes quick disconnect buffer exchange ports, safety lid with retractable sheathed leads, UVT gel tray with gasketed end gates, two 25 well and two 50 well combs (1.5 mm thick).