LOCTITE® GRD C (084) Thread lockers, Henkel Corporation
Supplier: Loctite
195776
76313-916EA 99.35 USD
76313-916
LOCTITE® GRD C (084) Thread lockers, Henkel Corporation
Adhesives
LOCTITE® Letter Grade C (084) Thread lockers are used to lock and seal fine threaded nuts, bolts, and studs in a wide variety of applications.
  • Medium strength
These are removable with tools.
Ordering information: Ten pieces per case. Must be ordered in case quantity.
Note: This is a regional approval. Please contact a local Technical Service Center for more information and clarification.
Packaging: 50 ml bottle, 0.077 kg Package Weight, 5.9L×2.9W×15.9H cm Packaging Dimension.