LOCTITE® 640 Retaining Compounds, Henkel Corporation
Supplier: Loctite
135520
76313-910EA 7.31 USD
76313-910
LOCTITE® 640 Retaining Compounds, Henkel Corporation
Adhesives
Ideal for parts requiring a longer positioning time.
  • Green in color
  • Low viscosity
  • High strength
These are methacrylate-ester based retaining compound. Fixturing time on steel is 2 hours. Also used for active metals such as brass. This is a high strength product with good temperature resistance. Maximum diametrical clearance is 0.1 mm, and service temperature is –55 to 175 °C. Fluorescence allows product presence to be monitored.
Ordering information: Ten pieces per case. Must be ordered in case quantity.
Packaging: 50 ml bottle, 0.081 kg Package Weight, 5.9L×2.9W×15.9H cm Packaging Dimension.