Luer Slip Syringes, B. Braun, B Braun
Supplier: B Braun
9166017V-02
76313-880CS 1577.79 USD
76313-880
Luer Slip Syringes, B. Braun, B Braun
Syringes General-Purpose Syringes
Manufactured only from laboratory polypropylene and polyethylene, these unique plastic syringes have a positive safety stop to help prevent accidental spills.
  • Ideal for simple injections, vaccinations and more
  • Compatible with all needles
  • Green plunger for easy recognition of filling volumes
  • Smooth, steady operation
  • Also available with Sterican® needles for unmatched sharpness and patient comfort
Injekt™ syringes do not contain rubber tree latex, silicone oil, styrene, or DEHP. Sterilization method: EO. Total shelf life: 60 months.