SW® GraphEx® ANSI Level A4 Cut-Resistant and 6 Abrasion-Resistant Gloves with AxiFybr® Technology, SW Safety Solutions
Supplier: SW Safety Solutions
G462002 G462003 G462006
76313-854BG 173.84 USD
76313-854 76313-856 76313-858 76313-860 76313-862
SW® GraphEx® ANSI Level A4 Cut-Resistant and 6 Abrasion-Resistant Gloves with AxiFybr® Technology, SW Safety Solutions
Gloves
Proprietary AxiFybr™ yarn is engineered at the molecular level enabling GraphEx® products to provide the highest level of cut protection with improved comfort, fit, and flexibility.
  • Even at advanced abrasion levels, these are touch-screen compatible
  • Proprietary yarn technology is 45% stronger and 21% more cut-resistant without sacrificing fit or comfort
  • Hi-Vis cuff provides instant indication of specific cut level and size from a distance
SW GraphEx™ cut resistant gloves with AxiFybr™ introduces a new proprietary yarn, engineered at the molecular level, enabling GraphEx™ gloves to provide the highest level of cut protection without loss of comfort, fit, and flexibility. Because of the AxiFybr™ high cut resistance, GraphEx™ gloves are more dexterous than similar rated gloves, allowing for greater protection without hand fatigue.
GraphEx™ liners are prewashed and form fitted to reduce strain on hands and allow for exceptional breathability. These gloves are ¹/₄ dipped in black micro porous nitrile.
This palm and finger coating provides high abrasion resistance that extends the use of the gloves by two-fold. The nitirle coating also offers superior touchscreen sensitivity.
Certifications: WBENC Women Owned. NCMSDC Minority Business. Small Business Enterprise. SHA Dermatologically Approved; EAC TP TC 019/2011; CE 0493; CE EN 455.
Ordering information: Glove sizes are as follows; S (7), M (8), L (9), XL (10), XXL (11).