Presston™ 1000 Positive Pressure Manifold 96-Well Plate, Agela Technologies
Supplier: Phenomenex
AH1-7033
76313-842EA 10947.22 USD
76313-842
Presston™ 1000 Positive Pressure Manifold 96-Well Plate, Agela Technologies
Manifolds
Streamline 96-well plate processing for easier sample preparation with a pneumatic positive pressure manifold.
  • Pneumatic handling
  • Consistent flow rates
  • Uniform flow rates for better reproducibility
  • Safe and easy-to-use
  • Ensure reliability
Presston™ 1000 positive pressure manifold for 96-well plate sample preparation processing offers an advanced alternative to traditional vacuum manifold processing. It applies pressure from above to pass liquids through the sample preparation product at a constant flow. To offer a wider applicability, it is equipped for processing samples in both a low pressure setting for typical solid phase extraction steps and a high pressure setting for very viscous sample processing and for sample dry down. In addition, it is compatible with standard 96-well plates, microelution plates, and 1 ml tubes with the addition of a tabless tube holder.