Dextrose 5% and 0.45% Sodium Chloride for injection

Supplier: B BRAUN MEDICAL

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

L6121
76313-650CS 143.82 USD
76313-650
Dextrose 5% and 0.45% Sodium Chloride for injection
D(+)-Glucose

Dextrose and Sodium Chloride Injections USP are sterile, nonpyrogenic and contain no bacteriostatic or antimicrobial agents. These products are intended for intravenous administration.


These intravenous solutions are indicated for use in adults and pediatric patients as sources of calories and water for hydration.


Not made with latex, PVC or DEHP.


Specific Gravity: 1.020


NDC number: 00264-7612-10

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
MDL Number: MFCD00148912
CAS Number: 50-99-7
Merck Index: 14,04459

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR