Ringer lactate solution USP for irrigation
Supplier: B BRAUN MEDICAL
R8306
76313-490CS 308.43 USD
76313-490
Ringer lactate solution USP for irrigation
Ringer lactate solution
Lactated Ringer’s Irrigation is useful as an irrigating fluid for body joints because it approximates the electrolyte composition of synovial fluid, and provides a transparent fluid medium with optical properties suitable for good visualization of the interior joint surface during endoscopic examination. During arthroscopic surgical procedures, the solution acts as a lavage for removing blood, tissue fragments, and bone fragments.
  • NDC number: 00264-7389-60
Lactated Ringer’s Irrigation is indicated for use as an arthroscopic irrigating fluid with endoscopic instruments during arthroscopic procedures requiring distension and irrigation of the knee, shoulder, elbow, or other bone joints.
Not made with natural rubber latex, DEHP or PVC.