Ringer lactate solution USP for injection
Supplier: B BRAUN MEDICAL
L7501
76313-478CS 381.41 USD
76313-478
Ringer lactate solution USP for injection
Ringer lactate solution
Lactated Ringer's Injection USP is sterile, nonpyrogenic and contains no bacteriostatic or antimicrobial agents. This product is intended for intravenous administration in a single dose container.
  • NDC number: 00264-7750-10
This solution is indicated for use in adults and pediatric patients as a source of electrolytes and water for hydration.
Not made with latex, PVC or DEHP.
Specific Gravity: 1.006