Qscript lyo 1-step, Quantabio
Supplier: Quantabio
95198-024
76312-730EA 100 USD
76312-730
Qscript lyo 1-step, Quantabio
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Qscript lyo 1-step is a lyophilized single-reaction reagent optimized for highly sensitive and reproducible one-step RT-qPCR using hydrolysis probes.
  • Dry, stable, easy and better 1-step RT-qPCR
  • Lyophilized single tube format easy to use, reduces cross-contamination
  • High sensitivity and specificity, detect as low as 0.5 pg RNA
  • Wide dynamic range from 0.5 to 500 pg RNA
  • Superior multiplexing, Plex up to 5 targets per reaction
  • Eliminate freezer storage up to 9 months stability room temperature
The reagent contains a hot-start thermo-stable polymerase, a genetically engineered reverse transcriptase as well as other components to ensure higher performance detection of up to 5 targets with maximum sensitivity and specificity. The enhanced stability of the freeze-dried master mix enables convenient shipping and storage at room temperature. The single tube reaction facilitates easy reaction set up while preventing potential cross-contamination.