Whatman™ Desiccant Pack, Whatman products (Cytiva)
Supplier: Cytiva
10548239
76312-378PK 29.92 USD
76312-378
Whatman™ Desiccant Pack, Whatman products (Cytiva)
Desiccators Desiccants
1 g packs of desiccant ensure that DNA sampling cards remain dry during transport or storage.
  • High vapor transmission rate
  • Substrate is made with Tyvek® material
  • Non-dusting, strong and hard to tear