Sterile Weighing Boats
Supplier: Heathrow Scientific
120710
76312-328PK 87.31 USD
76312-328
Sterile Weighing Boats
Boats Weighing Boats
Ideal for applications where there is concern of cross contamination and 'clean' is a necessity.
  • Made from 100% virgin anti-static polystyrene
  • Lot number traceability for quality assurance
  • Bend flexible boats at points to form pour spouts
  • Flat bottoms and sloping sides provide accurate transfer
  • Temperature range: 10 to 70 °C (14 to 158 °F)
  • Can be used with microwave moisture analyzers
Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.