KIMBLE® KimCote® GLS 80® Media Bottles, Clear, Wide Mouth, with Blue Polypropylene Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
14390-0250 14390-0500 14390-1000 14390-2000 14390-5000
76312-298CS 242.35 USD
76312-298 76312-300 76312-302 76312-304 76312-306
KIMBLE® KimCote® GLS 80® Media Bottles, Clear, Wide Mouth, with Blue Polypropylene Caps, DWK Life Sciences
Bottles Glass Bottles
Get the versatility, purity, and durability needed to achieve results for demanding laboratory storage, bioprocess, and production applications.
  • KimCote® coating provides safety and reduces hazards of shattered glass
  • Blue, quick-release, linerless polypropylene closure and pour ring are rated up to 140 °C
  • Autoclavable
Ideal for storage and transport of high-value sample substances, with a wide-mouth design that provides ease of use for filling and dispensing of powders, viscous liquids, and slurries. Durable, fired on ceramic ink graduations and marking patch withstand the rigors of daily laboratory, bioprocess, and production use. KIMBLE® 3.3 borosilicate glass is low-extractable, USP Type 1, with a full temperature and chemical resistance range.