KIMBLE® GLS 80® Media Bottles, Clear, Wide Mouth, with Blue Polypropylene Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
14393-0500 14393-1000 14393-2000 14393-5000 14393-10L 14393-20L 14393-0250
76312-272CS 90.99 USD
76312-272 76312-274 76312-276 76312-278 76312-280 76312-282 76312-318
KIMBLE® GLS 80® Media Bottles, Clear, Wide Mouth, with Blue Polypropylene Caps, DWK Life Sciences
Bottles Glass Bottles
Get the versatility, purity, and durability needed to achieve results for demanding laboratory storage, bioprocess, and production applications.
  • Wide-mouth design
  • Kimax® 3.3 borosilicate glass, USP Type 1
  • Autoclavable
Ideal for storage and transport of high-value sample substances, with a wide-mouth design that provides ease of use for filling and dispensing of powders, viscous liquids, and slurries. Durable, fired on ceramic ink graduations and marking patch withstand the rigors of daily laboratory, bioprocess, and production use. Glass is low-extractable, with a full temperature and chemical resistance range. Complete with blue, quick-release, linerless polypropylene closure and pour ring. Rated up to to 140 °C.