Versaflo™ Powered Air Purifying Respirator (PAPR) Assembly, 3M™
Supplier: 3M
7100035758
76312-266CS 1717.88 USD
76312-266
Versaflo™ Powered Air Purifying Respirator (PAPR) Assembly, 3M™
Respirators Respiratory Systems
TR-614N assembly may be combined with 3M™ Versaflo™ branded headtops and filters or cartridges to create a complete, compact, lightweight, easy to use PAPR for protection against certain particulates, vapors, and gases.
  • Easy to use
  • Easy to buy assembly of commonly used components
  • Lightweight design
  • Easy to clean
  • Hazard type: Silica
Single and four bay chargers are available to charge the battery included in the TR-614N assembly. The TR-614N assembly may be the prefered choice of customers working in dusty environments such as pharmaceutical, dry food, or dry cosmetic manufacturing.
Battery Life: 5.5 to 13 hours
Certifications: NIOSH approved.
Ordering information: The TR-614N assembly includes motor/blower (TR-602N), standard battery (TR-630), length adjusting breathing tube (BT-30), easy clean belt (TR-627) and airflow indicator (TR-971). Headtops, filters, cartridges, and battery charger are all sold separately.