Phenol Red Broth with Dulcitol, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Y305
76311-618PK 777.04 USD
76311-618
Phenol Red Broth with Dulcitol, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the differentiation of microorganisms by carbohydrate fermentation. Contains a durham tube for gas detection.