Memmert Reinforced Stainless Steel Shelves
Supplier: Memmert
B32550
76311-538EA 1206.75 USD
76311-538
Memmert Reinforced Stainless Steel Shelves
Furniture Shelves
Reinforced stainless steel grid shelves.
  • With guide bars and fixing screws
  • Requires option K1
Caution: Consider maximum loading of chamber; max. loading 60 kg