Notrax® 448 Autoclavable Bubble Floor Matting, Justrite®
Supplier: SUPERIOR MANUFACTURING GROUP INC
448S0023GY
76310-964EA 323.44 USD
76310-964
Notrax® 448 Autoclavable Bubble Floor Matting, Justrite®
Floor Mats Traction and Anti-Fatigue Floor Mats
This rubber anti-fatigue floor mat features a bubble top design.
  • Anti-microbial formula resists most Gram-positive and Gram-negative bacteria
  • Long-lasting rubber compound
  • Durable rubber top surface easy to wipe clean using most hospital grade cleaners or autoclaves
  • Beveled edges on all four sides
  • 6 sq. ft. coverage area
Offset bubbles on both the top and bottom of the mat provides flex and creates airflow resulting in maximum comfort for workers required to be on their feet for long periods of time. An industrial grade antimicrobial agent that inhibits the growth of most Gram-positive and Gram-negative bacteria and fungus is manufactured directly into the rubber compound. This ensures that the antimicrobial properties won’t wear-off over time like many post applied topical agents. Although intended for use in dry environments, the top surface design is easy to wipe clean and the durable rubber compound is highly resistant to many hospital grade cleaners and disinfectants so occasional spills can be cleaned up quickly and easily.