Notrax® 510 Diamond-Tuff™ Max Floor Matting, Justrite®
Supplier: NOTRAX USA, INC.
510R0036YB
76310-844EA 3562.81 USD
76310-844
Notrax® 510 Diamond-Tuff™ Max Floor Matting, Justrite®
Floor Mats Traction and Anti-Fatigue Floor Mats
Diamond-Tuff™ Max is a durable anti-fatigue/anti-slip mat with a vinyl diamond plate top surface that provides sure footing and a full 1" thick foam base for maximum comfort.
  • Durable diamond pattern vinyl laminate top surface for added traction
  • Dense closed cell PVC foam base provides added cushioning comfort
  • Beveled edges on all 4 sides
  • 225 sq. ft. coverage area
It is available in both workstation sizes and full rolls for applications that require a longer mat such as assembly lines. The non-directional diamond plate top surface is easy to sweep or mop clean.
Material Sponge/Vinyl