Anti-ATF6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-GU9 600-401-GU9S
76310-446EA 551.64 USD
76310-446 76310-448
Anti-ATF6 Rabbit Polyclonal Antibody
Antibodies
ATF6 belongs to the bZIP (basic-leucine zipper domain transcription factor) family. Activating target genes for the unfolded protein response during endoplasmic reticulum stress. Embeded in the endoplasmic reticulum, it functions as an ER stress sensor/stress transducer. Following stress-induced proteolysis it functions as a nuclear transcription factor via a cis-acting ER stress response element (ERSE) that is present in the promoters of genes encoding ER chaperones. Binds DNA on the 5'-CCAC[GA]-3' half of the ER stress response element (ERSE) (5'-CCAAT-N(9)-CCAC[GA]-3') and of ERSE II (5'-ATTGG-N-CCACG-3'). Binding to ERSE requires binding of NF-Y to ERSE. ATF6 could also be involved in the activation of transcription by the serum response factor. This protein may play a role in the development of the foveal and cone function in the retina. Therefore, attributing to disorders such as achromatopsia, blindness, nystagmus, myopia, cataracts and more. Anti-ATF6 Antibody is useful for researchers studying eye development, protein folding and processing, Endoplasmic Reticulum Stress Pathways, Glucose and Energy Metabolism Research.

Type: Primary
Antigen: ATF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR