Anti-TLR4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-MK3 600-401-MK3S
76309-356EA 551.64 USD
76309-356 76309-358
Anti-TLR4 Rabbit Polyclonal Antibody
Antibodies
TLR4 (Toll-Like Receptor 4). Toll-like receptors (TLRs) are signaling molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses.  These proteins act through adaptor molecules such as MyD88 and TIRAP to activate various kinases and transcription factors such as Protein Kinase C (PKC) alpha/beta and NF-kB.  Studies with TLR4-deficient mice indicate that the main ligand for TLR is lipopolysaccharide.  Consequently, these mice also showed increased susceptibility to Gram-negative sepsis. Mutations in this gene have been associated with differences in LPS responsiveness, macular degeneration, and pertussis. Anti-TLR4 Antibody is useful for researchers interested in transcription factor research, cytokine research, MIF action through Endocytic Pathway, and rheumatoid arthritis.

Type: Primary
Antigen: TLR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR