VWR® Digital General Purpose Water Baths
Supplier: VWR International
76308-830 76308-834 76308-896 76308-904
76308-830EA 704.44 USD
76308-830 76308-834 76308-896 76308-904
VWR® Digital General Purpose Water Baths
Baths Water Baths, Unstirred
These premium digital general-purpose water baths are designed with high-gabled lids with hinge to hold lid up and allow condensate to drain into bath.
  • Easy-to-read, colour, touch screen
  • Five pre-sets for frequently used temperatures
  • Large buttons for easy navigation
  • Eco mode for heat-off use
  • Calibration offset feature allow user to adjust the temperature display read out at any one specific point
  • Safety thermostat and alarms
  • 2 year warranty
Features five language (Chinese, English, French, German and Spanish) setting option and have rear-mounted drain on 10, 20 and 28 L models.
Certifications: cETLus compliant.