VWR® Upholstered Lab Stools
Supplier: VWR International
76308-780 76308-784 76308-788 76308-792
76308-780EA 576.1 USD
76308-780 76308-784 76308-788 76308-792
VWR® Upholstered Lab Stools
Chairs
This VWR® upholstered lab stool features a roomy and comfortable 16" Ø seat with 3.5" of resilient foam cushioning and adjustability proven to provide reliable ergonomic performance, shift after shift, year after year.
  • Exclusive black microdot-textured vinyl upholstery combines practical performance with a sophisticated look
  • Other components include an easy-to-use ergonomic task control with seat height and back tilt adjustments and 26" Ø black composite base
  • Designed for laboratory applications, including those in healthcare, education and industrial
More Product Information